Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Asb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236158.2 | XP_006236220.1 | 9 | 1849 | ||
XM_008762831.1 | XP_008761053.1 | 9 | 2005 | |||
XM_008762832.1 | XP_008761054.1 | 9 | 1806 | |||
XM_008762833.1 | XP_008761055.1 | 9 | 2027 |