Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ublcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014117.1 | NP_001014139.1 | 7 | 650 | ||
XM_006246140.1 | XP_006246202.1 | 7 | 680 | |||
GenBank mRNA | BC089210.1 | - | 7 | 650 | 7302690 | |
FQ233702.1 | - | 7 | 653 |