Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108276.1 | NP_001101746.1 | 2 | 571 | ||
XM_006246505.2 | XP_006246567.1 | 2 | 571 | |||
GenBank mRNA | BC160894.1 | - | 2 | 522 | 7446534 |