Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108278.1 | NP_001101748.1 | 6 | 560 | ||
XM_006246743.1 | XP_006246805.1 | 7 | 681 | |||
XM_006246744.2 | XP_006246806.1 | 7 | 582 | |||
XM_008767837.1 | XP_008766059.1 | 6 | 597 | |||
GenBank mRNA | FQ218736.1 | - | 7 | 541 |