Gene Symbol: Tada2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012141.1 | NP_001012141.1 | 4 | 337 | ||
XM_006247042.2 | XP_006247104.1 | 3 | 246 | |||
GenBank mRNA | BC079084.1 | - | 4 | 337 | 7123007 | |
EF121983.1 | - | 3 | 176 | |||
EF121984.1 | - | 3 | 176 | |||
EF121985.1 | - | 3 | 176 |