Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108300.1 | NP_001101770.1 | 2 | 230 | ||
XM_006247379.2 | XP_006247441.1 | 3 | 351 | |||
XM_006247380.2 | XP_006247442.1 | 3 | 351 |