Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007725.1 | NP_001007726.1 | 5 | 590 | ||
XM_006247624.2 | XP_006247686.1 | 5 | 600 | |||
XM_006247627.2 | XP_006247689.1 | 3 | 434 | |||
XM_008768377.1 | XP_008766599.1 | 4 | 1197 | |||
XM_008768378.1 | XP_008766600.1 | 6 | 649 | |||
XM_008768379.1 | XP_008766601.1 | 4 | 509 | |||
GenBank mRNA | BC079465.1 | - | 5 | 590 | 7122370 | |
FQ220624.1 | - | 2 | 296 | |||
FQ231566.1 | - | 5 | 458 | |||
FQ231887.1 | - | 5 | 458 |