Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033274.4 | NP_150377.1 | 3, 4 | 375 | ||
XM_005266003.2 | XP_005266060.1 | 3, 4 | 327 | |||
GenBank mRNA | AF311317.1 | - | 3, 4 | 284 | ||
AF326918.1 | - | 3, 4 | 296 | |||
AK091464.1 | - | 3, 4 | 327 | |||
AK308182.1 | - | 3, 4 | 327 | |||
HY088077.1 | - | 3, 4 | 375 | |||
Y13786.2 | - | 3, 4 | 325 |