Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012148.1 | NP_001012148.1 | 10 | 1181 | ||
XM_006249461.1 | XP_006249523.1 | 10 | 1721 | |||
XM_006249462.2 | XP_006249524.1 | 10 | 1065 | |||
GenBank mRNA | BC085774.1 | - | 10 | 1181 | 7096477 |