Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014132.1 | NP_001014154.1 | 11, 12 | 1288 | ||
XM_008769865.1 | XP_008768087.1 | 10, 11 | 1953 | |||
XM_008769866.1 | XP_008768088.1 | 10, 11 | 1953 | |||
GenBank mRNA | BC089843.1 | - | 11, 12 | 1288 | 7368358 |