Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001181.4 | NP_001172.1 | 8 | 1026 | ||
NM_001201352.1 | NP_001188281.1 | 8 | 1011 | |||
NM_080912.3 | NP_550434.1 | 8 | 1114 | |||
NM_080913.3 | NP_550435.1 | 8 | 954 | |||
NM_080914.2 | NP_550436.1 | 7 | 772 | |||
XM_005256648.2 | XP_005256705.1 | 9 | 1064 | |||
XM_006721524.2 | XP_006721587.2 | 7 | 831 | |||
XM_006721526.1 | XP_006721589.1 | 9 | 1511 | |||
XM_011523862.2 | XP_011522164.2 | 9 | 1117 | |||
XM_011523863.2 | XP_011522165.1 | 8 | 986 | |||
XM_011523864.2 | XP_011522166.1 | 8 | 1021 | |||
XM_011523865.1 | XP_011522167.1 | 9 | 1547 | |||
XM_011523866.1 | XP_011522168.1 | 9 | 1097 | |||
XM_017024651.1 | XP_016880140.1 | 8 | 965 | |||
XM_017024652.1 | XP_016880141.1 | 9 | 1495 | |||
XM_017024653.1 | XP_016880142.1 | 9 | 1583 | |||
GenBank mRNA | AF529374.1 | - | 7 | 744 | ||
BC017251.2 | - | 8 | 902 | 5087419 | ||
M11025.1 | - | 8 | 879 | |||
U97197.1 | - | 7 | 702 | |||
X55283.1 | - | 8 | 877 | |||
X55284.1 | - | 8 | 956 |