Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317061.1 | NP_001303990.1 | 13 | 2254 | ||
NM_003804.4 | NP_003795.2 | 11 | 1976 | |||
XM_005249457.3 | XP_005249514.2 | 11 | 1899 | |||
XM_017011403.1 | XP_016866892.1 | 8 | 2008 | |||
XM_017011404.1 | XP_016866893.1 | 5 | 1216 | |||
GenBank mRNA | AB208926.1 | - | 13 | 2163 | ||
AK096523.1 | - | 5 | 744 | |||
AK304701.1 | - | 9 | 1565 | |||
AK314176.1 | - | 11 | 1976 | |||
BC126254.1 | - | 10 | 1770 | 8991970 | ||
BC126256.1 | - | 10 | 1770 | 8991972 | ||
BX348786.2 | - | 4 | 827 | |||
U25994.1 | - | 5 | 847 | |||
U50062.1 | - | 10 | 1744 |