Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251033.2 | XP_006251095.1 | 20 | 5187 | ||
XM_341223.7 | XP_341224.5 | 20 | 2983 |