Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TNFSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190942.1 | NP_001177871.1 | 3 | 515 | ||
NM_003810.3 | NP_003801.1 | 5 | 663 | |||
NR_033994.1 | 4 | 620 | ||||
GenBank mRNA | AK296085.1 | - | 3 | 483 | ||
AK312742.1 | - | 5 | 631 | |||
BC009795.1 | - | 5 | 560 | |||
BC032722.1 | - | 5 | 615 | 5504670 | ||
BT019564.1 | - | 5 | 540 | |||
CR456895.1 | - | 5 | 540 | |||
EU183231.1 | - | 3 | 392 | |||
U37518.1 | - | 5 | 627 | |||
U57059.1 | - | 5 | 630 |