Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198774.2 | NP_942069.1 | 2, 3 | 306 | ||
GenBank mRNA | AY283801.1 | - | 2, 3 | 329 | ||
BC088092.1 | - | 2, 3 | 306 | 7368117 |