Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024795.1 | NP_001019966.1 | 6, 7 | 1020 | ||
XM_006253633.2 | XP_006253695.1 | 6, 7 | 2017 | |||
XM_006253634.2 | XP_006253696.1 | 6, 7 | 1010 | |||
XM_006253637.2 | XP_006253699.1 | 6, 7 | 2539 | |||
XM_006253641.2 | XP_006253703.1 | 6, 7 | 1953 | |||
XM_006253642.2 | XP_006253704.1 | 6, 7 | 2475 | |||
XM_006253644.2 | XP_006253706.1 | 6, 7 | 946 | |||
XM_008771498.1 | XP_008769720.1 | 6, 7 | 2536 | |||
GenBank mRNA | BC098030.1 | - | 6, 7 | 1020 | 7456296 |