Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254677.2 | XP_006254739.1 | 9 | 1497 | ||
XM_006254680.2 | XP_006254742.1 | 8 | 1203 | |||
XM_008772179.1 | XP_008770401.1 | 9 | 1497 |