Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025722.2 | NP_001020893.2 | 7 | 1025 | ||
NM_001109936.1 | NP_001103406.1 | 8 | 574 | |||
NM_001109937.1 | NP_001103407.1 | 8 | 535 | |||
GenBank mRNA | BC098804.1 | - | 7 | 1025 | 7376321 | |
DN932869.1 | - | 8 | 570 | |||
DN935585.1 | - | 8 | 594 |