Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108466.1 | NP_001101936.1 | 2 | 1132 | ||
XM_006228268.2 | XP_006228330.1 | 3 | 1082 | |||
XM_006228269.2 | XP_006228331.1 | 3 | 1079 | |||
XM_006228270.1 | XP_006228332.1 | 3 | 1033 | |||
XM_006228271.1 | XP_006228333.1 | 3 | 1030 | |||
XM_006228272.2 | XP_006228334.1 | 3 | 1386 | |||
XM_006228273.2 | XP_006228335.1 | 3 | 1383 |