Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127552.1 | NP_001121024.1 | 8 | 1197 | ||
XM_006228856.2 | XP_006228918.1 | 8 | 1245 | |||
XM_006228857.2 | XP_006228919.1 | 8 | 1245 | |||
XM_006228858.2 | XP_006228920.1 | 7 | 1068 | |||
GenBank mRNA | BC166537.1 | - | 8 | 1197 | 7191211 |