Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003842.4 | NP_003833.4 | 9 | 1437 | ||
NM_147187.2 | NP_671716.2 | 10 | 1350 | |||
NR_027140.1 | 9 | 1244 | ||||
XM_017013944.1 | XP_016869433.1 | 9 | 1225 | |||
GenBank mRNA | AB451272.1 | - | 9 | 1144 | ||
AB451399.1 | - | 9 | 1144 | |||
AF012535.1 | - | 10 | 1196 | |||
AF012628.1 | - | 10 | 1057 | |||
AF016266.1 | - | 9 | 1261 | |||
AF016268.1 | - | 10 | 1166 | |||
AF016849.1 | - | 9 | 1288 | |||
AF018657.1 | - | 10 | 1057 | |||
AF018658.1 | - | 9 | 1144 | |||
AF020501.1 | - | 10 | 1057 | |||
AF022386.1 | - | 10 | 1057 | |||
AF153687.1 | - | 10 | 1175 | |||
AF192548.1 | - | 10 | 1157 | |||
AK291216.1 | - | 9 | 1264 | |||
AK296128.1 | - | 5 | 450 | |||
AK298602.1 | - | 9 | 1088 | |||
AY358277.1 | - | 10 | 1196 | |||
BC001281.1 | - | 9 | 1291 | 3458466 | ||
BX537617.1 | - | 4 | 542 | |||
BX538104.1 | - | 9 | 1266 | |||
BX538179.1 | - | 4 | 542 | |||
CR541898.1 | - | 9 | 1144 | |||
CX870630.1 | - | 5 | 649 | |||
FJ200482.1 | - | 8 | 1038 |