Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100736.1 | NP_001094206.1 | 1 | 482 | ||
GenBank mRNA | BC169098.1 | - | 1 | 482 | 7939241 |