Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwwp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108507.2 | NP_001101977.1 | 2 | 795 | ||
XM_006230466.2 | XP_006230528.1 | 2 | 3164 | |||
XM_006230469.2 | XP_006230531.1 | 2 | 3163 | |||
GenBank mRNA | BC162015.1 | - | 2 | 795 | 9092167 |