Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760189.1 | XP_008758411.1 | 2 | 399 | ||
XM_341975.7 | XP_341976.4 | 2 | 399 |