Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108515.1 | NP_001101985.1 | 17 | 3085 | ||
XM_006230843.2 | XP_006230905.1 | 17 | 2857 | |||
XM_006230844.2 | XP_006230906.1 | 17 | 3456 | |||
XM_008760152.1 | XP_008758374.1 | 16 | 3105 |