Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014165.1 | NP_001014187.1 | 2 | 219 | ||
XM_006231237.2 | XP_006231299.1 | 2 | 178 | |||
XM_006231239.2 | XP_006231301.1 | 2 | 95 | |||
XM_006231240.2 | XP_006231302.1 | 1 | 65 | |||
XM_008760342.1 | XP_008758564.1 | 3 | 248 | |||
XM_008760343.1 | XP_008758565.1 | 3 | 249 | |||
GenBank mRNA | BC079024.1 | - | 2 | 219 | 7113918 |