Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212507.2 | NP_997672.1 | 2 | 443 | ||
GenBank mRNA | BC088208.1 | - | 2 | 443 | 7381953 | |
FQ227971.1 | - | 2 | 397 | |||
FQ235217.1 | - | 2 | 368 |
Gene Symbol: LOC103694381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772776.1 | XP_008770998.1 | 2 | 738 |