Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025726.1 | NP_001020897.1 | 4 | 369 | ||
XM_008772734.1 | XP_008770956.1 | 4 | 403 | |||
GenBank mRNA | BC099089.1 | - | 4 | 369 | 7456350 |