Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003863.3 | NP_003854.1 | 4 | 1060 | ||
GenBank mRNA | AF061729.1 | - | 5 | 424 | ||
BC015233.1 | - | 4 | 1060 | 3901944 | ||
BC015374.1 | - | 2 | 1180 | 4181887 | ||
BC107863.1 | - | 4 | 434 | 6450798 |