Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003863.3 | NP_003854.1 | 4 | 878 | ||
GenBank mRNA | AB013361.1 | - | 4 | 250 | ||
CR542134.1 | - | 3 | 214 |