Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr89b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139486.1 | NP_001132958.1 | 4 | 379 | ||
XM_006232976.2 | XP_006233038.1 | 2 | 368 | |||
GenBank mRNA | AY724489.1 | - | 4 | 310 | ||
DY548897.1 | - | 4 | 379 |
Set Membership: |
![]() |