Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108567.2 | NP_001102037.1 | 5 | 341 | ||
XM_006233314.2 | XP_006233376.1 | 5 | 506 | |||
XM_006233315.1 | XP_006233377.1 | 4 | 329 | |||
GenBank mRNA | BC167113.1 | - | 5 | 341 | 6889959 | |
FQ211292.1 | - | 5 | 344 | |||
FQ211369.1 | - | 5 | 343 | |||
FQ216454.1 | - | 702 | ||||
FQ221198.1 | - | 5 | 343 | |||
FQ221629.1 | - | 5 | 343 | |||
FQ221933.1 | - | 5 | 343 | |||
FQ223193.1 | - | 5 | 343 | |||
FQ224761.1 | - | 5 | 349 | |||
FQ229743.1 | - | 5 | 343 |