Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108620.1 | NP_001102090.1 | 31 | 4484 | ||
XM_008762795.1 | XP_008761017.1 | 31 | 4661 | |||
XM_008762796.1 | XP_008761018.1 | 31 | 4662 | |||
XM_008762797.1 | XP_008761019.1 | 30 | 4341 | |||
XM_008762798.1 | XP_008761020.1 | 30 | 4241 |