Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100750.1 | NP_001094220.1 | 8 | 805 | ||
GenBank mRNA | BC079367.1 | - | 8 | 798 | ||
BC089863.1 | - | 7 | 663 | |||
BC099746.1 | - | 8 | 789 | |||
BC160886.1 | - | 8 | 805 | 7385389 |