Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tstd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108663.1 | NP_001102133.1 | 8 | 1238 | ||
XM_006238084.1 | XP_006238146.1 | 8 | 1160 | |||
XM_006238085.2 | XP_006238147.1 | 8 | 1317 | |||
XM_006238086.1 | XP_006238148.1 | 7 | 1053 | |||
XM_006238088.2 | XP_006238150.1 | 8 | 1160 | |||
XM_006238089.2 | XP_006238151.1 | 8 | 1160 | |||
XM_006238090.2 | XP_006238152.1 | 8 | 1160 | |||
XM_008763698.1 | XP_008761920.1 | 8 | 1160 |