Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103689941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764128.1 | XP_008762350.1 | 15 | 2379 |
Gene Symbol: Inpp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100755.1 | NP_001094225.1 | 20 | 2662 | ||
GenBank mRNA | BC098852.1 | - | 1809 |