Search Thermo Fisher Scientific
Gene Symbol: Tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024800.1 | NP_001019971.1 | 4 | 411 | ||
XM_006240178.2 | XP_006240240.1 | 4 | 1634 | |||
GenBank mRNA | BC094308.1 | - | 4 | 411 | 7316392 |