Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108720.1 | NP_001102190.1 | 2 | 828 | ||
XM_008764818.1 | XP_008763040.1 | 2 | 863 | |||
XM_008764819.1 | XP_008763041.1 | 3 | 1005 | |||
XM_008764820.1 | XP_008763042.1 | 2 | 864 | |||
XM_008764821.1 | XP_008763043.1 | 2 | 864 |