Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047915.1 | NP_001041380.1 | 1 | 189 | ||
GenBank mRNA | AY390379.1 | - | 1 | 189 |