Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108739.1 | NP_001102209.1 | 3 | 447 | ||
XM_006241164.2 | XP_006241226.1 | 3 | 480 | |||
XM_008765244.1 | XP_008763466.1 | 3 | 1070 | |||
GenBank mRNA | BC166979.1 | - | 3 | 350 | 7315264 | |
FQ218051.1 | - | 1 | 257 |