Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 13 | 1626 | ||
NM_153050.2 | NP_694690.1 | 13 | 1626 | |||
NM_153051.2 | NP_694691.1 | 13 | 1626 | |||
XM_005261803.2 | XP_005261860.1 | 14 | 1744 | |||
XM_005261804.3 | XP_005261861.1 | 13 | 1661 | |||
XM_005261805.2 | XP_005261862.1 | 14 | 1744 | |||
XM_005261806.2 | XP_005261863.1 | 13 | 1626 | |||
XM_005261808.2 | XP_005261865.1 | 14 | 1744 | |||
XM_017029028.1 | XP_016884517.1 | 12 | 1518 | |||
XM_017029029.1 | XP_016884518.1 | 12 | 1508 | |||
XM_017029030.1 | XP_016884519.1 | 12 | 1524 | |||
XM_017029031.1 | XP_016884520.1 | 12 | 1512 | |||
XM_017029032.1 | XP_016884521.1 | 13 | 1509 | |||
XM_017029033.1 | XP_016884522.1 | 7 | 945 | |||
XM_017029034.1 | XP_016884523.1 | 14 | 1720 | |||
GenBank mRNA | AB002369.1 | - | 13 | 1545 | ||
AF233436.2 | - | 10 | 1307 | |||
AF233437.1 | - | 10 | 1307 | |||
AF233438.1 | - | 10 | 1307 | |||
AK289581.1 | - | 13 | 1626 | |||
BC142713.1 | - | 13 | 1582 | 40148839 | ||
BC148216.1 | - | 12 | 1445 | 40082266 | ||
BC151217.1 | - | 13 | 1640 | 40148098 | ||
BC152455.1 | - | 13 | 1545 | 8862588 | ||
CR456525.1 | - | 11 | 1322 |