Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013221.1 | NP_001013239.1 | 6, 7 | 756 | ||
XM_006242226.2 | XP_006242288.1 | 6, 7 | 838 | |||
GenBank mRNA | BC083624.1 | - | 6, 7 | 756 | 7134121 |