Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRPF4B SIRNA Assays ›
Gene Symbol:
PRPF4B
Gene Name:
pre-mRNA processing factor 4B
Gene Aliases:
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1
Chromosome Location:
Chr.6: 4019402 - 4064983 on Build GRCh38
UniGene ID:
Hs.159014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17017
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRPF4B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003913.4
NP_003904.3
2
983
XM_011514970.2
XP_011513272.1
2
2916
XM_017011410.1
XP_016866899.1
2
2916
XM_017011411.1
XP_016866900.1
2
2916
XM_017011412.1
XP_016866901.1
2
2916
XM_017011413.1
XP_016866902.1
2
2916
XM_017011414.1
XP_016866903.1
2
2916
XM_017011415.1
XP_016866904.1
2
2916
XM_017011416.1
XP_016866905.1
2
2916
XM_017011417.1
XP_016866906.1
2
2916
XM_017011418.1
XP_016866907.1
2
2916
XM_017011419.1
XP_016866908.1
2
2916
XM_017011420.1
XP_016866909.1
2
2916
XM_017011421.1
XP_016866910.1
2
2916
XM_017011422.1
XP_016866911.1
2
2916
XM_017011423.1
XP_016866912.1
2
2916
XM_017011424.1
XP_016866913.1
2
2916
XM_017011425.1
XP_016866914.1
2
2916
XM_017011426.1
XP_016866915.1
2
2916
XM_017011427.1
XP_016866916.1
2
2916
XM_017011428.1
XP_016866917.1
2
2916
XM_017011429.1
XP_016866918.1
2
2916
XM_017011430.1
XP_016866919.1
2
2916
XM_017011431.1
XP_016866920.1
2
2916
XM_017011432.1
XP_016866921.1
2
2916
XM_017011433.1
XP_016866922.1
2
2916
XM_017011434.1
XP_016866923.1
2
2916
XM_017011435.1
XP_016866924.1
2
2916
GenBank mRNA
AB011108.1
-
2
956
AF283465.1
-
2
934
AK291244.1
-
2
961
AK303047.1
-
2
1008
AY029347.1
-
2
892
BC034969.1
-
2
996
4821304
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
protein phosphorylation
RNA splicing
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline