Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108773.1 | NP_001102243.1 | 3 | 812 | ||
XM_006243483.2 | XP_006243545.1 | 3 | 699 | |||
XM_006243485.2 | XP_006243547.1 | 3 | 738 | |||
XM_006243486.2 | XP_006243548.1 | 3 | 915 |