Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108774.1 | NP_001102244.1 | 3 | 243 | ||
XM_006243487.2 | XP_006243549.1 | 4 | 523 | |||
XM_006243488.2 | XP_006243550.1 | 3 | 378 | |||
XM_008766452.1 | XP_008764674.1 | 4 | 511 | |||
XM_008766453.1 | XP_008764675.1 | 3 | 411 |