Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1G SIRNA Assays ›
Gene Symbol:
CACNA1G
Gene Name:
calcium voltage-gated channel subunit alpha1 G
Gene Aliases:
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location:
Chr.17: 50560929 - 50629625 on Build GRCh38
UniGene ID:
Hs.591169
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17051
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256324.1
NP_001243253.1
9
2610
NM_001256325.1
NP_001243254.1
9
2610
NM_001256326.1
NP_001243255.1
9
2610
NM_001256327.1
NP_001243256.1
9
2610
NM_001256328.1
NP_001243257.1
9
2610
NM_001256329.1
NP_001243258.1
9
2610
NM_001256330.1
NP_001243259.1
9
2610
NM_001256331.1
NP_001243260.1
9
2610
NM_001256332.1
NP_001243261.1
9
2610
NM_001256333.1
NP_001243262.1
9
2610
NM_001256334.1
NP_001243263.1
9
2610
NM_001256359.1
NP_001243288.1
9
2610
NM_001256360.1
NP_001243289.1
9
2610
NM_001256361.1
NP_001243290.1
9
2610
NM_018896.4
NP_061496.2
9
2610
NM_198376.2
NP_938190.1
9
2610
NM_198377.2
NP_938191.2
9
2610
NM_198378.2
NP_938192.1
9
2610
NM_198379.2
NP_938193.1
9
2610
NM_198380.2
NP_938194.1
9
2610
NM_198382.2
NP_938196.1
9
2610
NM_198383.2
NP_938197.1
9
2610
NM_198384.2
NP_938198.1
9
2610
NM_198385.2
NP_938199.1
9
2610
NM_198386.2
NP_938200.1
9
2610
NM_198387.2
NP_938201.1
9
2610
NM_198388.2
NP_938202.1
9
2610
NM_198396.2
NP_938406.1
9
2610
NR_046054.1
9
2610
NR_046055.1
9
2610
NR_046056.1
9
2610
NR_046057.1
9
2610
NR_046058.1
9
2610
XM_006722160.3
XP_006722223.1
9
2749
XM_006722161.3
XP_006722224.1
9
2698
GenBank mRNA
AB012043.1
-
9
2246
AB032949.2
-
2
572
AF124351.1
-
9
2590
AF126965.1
-
9
2393
AF126966.1
-
9
2393
AF134985.1
-
9
2218
AF134986.1
-
9
2218
AF190860.1
-
9
2434
AF227744.1
-
9
2218
AF227745.1
-
9
2218
AF227746.1
-
9
2218
AF227747.1
-
9
2218
AF227748.1
-
9
2218
AF227749.1
-
9
2218
AF227750.1
-
9
2218
AF227751.1
-
9
2218
BC110995.1
-
9
2610
5502027
DQ494449.1
-
9
2218
DQ494450.1
-
9
2218
DQ494451.1
-
9
2218
DQ494452.1
-
9
2218
DQ494453.1
-
9
2218
DQ494454.1
-
9
2218
DQ494455.1
-
9
2218
DQ494456.1
-
9
2218
DQ494457.1
-
9
2218
DQ494458.1
-
9
2218
DQ494459.1
-
9
2218
DQ494460.1
-
9
2218
DQ494461.1
-
9
2218
DQ494462.1
-
9
2218
DQ494463.1
-
9
2218
DQ494464.1
-
9
2218
DQ494465.1
-
9
2218
DQ494466.1
-
9
2218
DQ494467.1
-
9
2218
DQ494468.1
-
9
2218
DQ494469.1
-
9
2218
DQ494470.1
-
9
2218
DQ494471.1
-
9
2218
DQ494472.1
-
9
2218
DQ494473.1
-
9
2218
DQ494474.1
-
9
2218
DQ494475.1
-
9
2218
DQ494476.1
-
9
2218
DQ494477.1
-
9
2218
DQ494478.1
-
9
2218
DQ494479.1
-
9
2218
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
response to nickel cation
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
SA node cell action potential
AV node cell action potential
SA node cell to atrial cardiac muscle cell signalling
AV node cell to bundle of His cell signaling
membrane depolarization during AV node cell action potential
membrane depolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
low voltage-gated calcium channel activity
voltage-gated calcium channel activity involved in AV node cell action potential
voltage-gated calcium channel activity involved SA node cell action potential
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0