Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177419.2 | NP_803154.1 | 19 | 2154 | ||
GenBank mRNA | AB066103.1 | - | 17 | 1911 | ||
BC061576.1 | - | 19 | 2157 | 6887693 |