Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003922.3 | NP_003913.3 | 22 | 4174 | ||
XM_017022699.1 | XP_016878188.1 | 22 | 4187 | |||
XM_017022700.1 | XP_016878189.1 | 22 | 4187 | |||
XM_017022701.1 | XP_016878190.1 | 22 | 4187 | |||
XM_017022702.1 | XP_016878191.1 | 22 | 4187 | |||
XM_017022703.1 | XP_016878192.1 | 22 | 4187 | |||
XM_017022704.1 | XP_016878193.1 | 22 | 4187 | |||
XM_017022705.1 | XP_016878194.1 | 22 | 4187 | |||
XM_017022706.1 | XP_016878195.1 | 22 | 4190 | |||
GenBank mRNA | AK226162.1 | - | 9 | 1237 | ||
AK296690.1 | - | 3 | 1120 | |||
U50078.1 | - | 22 | 4122 |