Gene Symbol: Bex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077435.1 | NP_001070903.1 | 3 | 744 | ||
GenBank mRNA | AY833555.1 | - | 3 | 744 |
Gene Symbol: Bex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY208292.1 | - | 721 |