Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109526.2 | NP_001102996.2 | 5, 6 | 468 | ||
XM_006245736.2 | XP_006245798.1 | 4, 5 | 774 | |||
XM_008767497.1 | XP_008765719.1 | 5, 6 | 1583 | |||
XM_008767498.1 | XP_008765720.1 | 3, 4 | 409 |